Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bruker Match
™
tubes
Products
Product Category
tubes (8)
Color
Manufacturer
Diameter (mm)
OD (mm)
Volume (µL)
Color
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
NORS175001
Bruker Match
™
tubes,
O.D. × L 1.7 mm × 100 mm, volume 45 μL
Expand
View Pricing
NORS255001
Bruker Match
™
tubes,
capacity 120 μL
Expand
View Pricing
NORS105001
Bruker Match
™
tubes,
O.D. × L 1 mm × 100 mm, volume 12 μL
Expand
View Pricing
NORS505001
Bruker Match
™
tubes,
O.D. × L 5.0 mm × 100 mm, volume 490 μL
Expand
View Pricing
NORS305001
Bruker Match
™
tubes,
O.D. × L 3.0 mm × 100 mm, volume 160 μL
Expand
View Pricing
NORS205001
Bruker Match
™
tubes,
O.D. × L 2.0 mm × 100 mm, volume 70 μL
Expand
View Pricing
NORS4255001
Bruker Match
™
tubes,
O.D. × L 4.25 mm × 100 mm, volume 370 μL
Expand
View Pricing
NORS405001
Bruker Match
™
tubes,
volume 310 μL
Expand
View Pricing