Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bromine
Synonym(s):
Bromine
Empirical Formula (Hill Notation):
Br
2
CAS Number:
7726-95-6
Molecular Weight:
159.81
EC Number:
231-778-1
Products
Product Category
salts (3)
acids (1)
Brand
Sigma-Aldrich (6)
Supelco (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Special Grade
ACS reagent (1)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
277576
Bromine,
ACS reagent, ≥99.5%
Expand
View Pricing
207888
Bromine,
reagent grade
Expand
View Pricing
8.20171
Bromine,
for synthesis
Expand
View Pricing
470864
Bromine,
≥99.99% trace metals basis
Expand
View Pricing
1.01948
Bromine,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
1.01945
Bromine,
extra pure
Expand
View Pricing
1.01947
Bromine,
99.9999 Suprapur
®
Expand
View Pricing
03-3360
Bromine,
JIS special grade, ≥99.0%
Expand
View Pricing
03-3370
Bromine,
SAJ first grade, ≥97.0%
Expand
View Pricing