Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Brij
®
L23
Synonym(s):
Brij 35, C
12
E
23
, Polyoxyethylene (23) lauryl ether
CAS Number:
9002-92-0
Products
Product Category
detergents (2)
Brand
Sigma-Aldrich (2)
Feature
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
16005
Brij
®
L23,
main component: tricosaethylene glycol dodecyl ether
Expand
View Pricing
P1254
Brij
®
L23,
suitable for Stein-Moore chromatography
Expand
View Pricing