Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Brass alloy, Cu70Zn30
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
GF66429119
Brass alloy, Cu70Zn30,
sphere, 2.0mm diameter, 5.08μm sphericity, precision sphere, grade 200, 100pcs
Expand
View Pricing
GF53394587
Brass alloy, Cu70Zn30,
200mm tube, 3.56mm outside diameter, 0.41mm wall thickness, 2.74mm inside diameter, as drawn
Expand
View Pricing
GF90397547
Brass alloy, Cu70Zn30,
1000mm tube, 3.56mm outside diameter, 0.41mm wall thickness, 2.74mm inside diameter, as drawn
Expand
View Pricing
GF34827150
Brass alloy, Cu70Zn30,
100mm tube, 3.56mm outside diameter, 0.41mm wall thickness, 2.74mm inside diameter, as drawn
Expand
View Pricing
GF74323188
Brass alloy, Cu70Zn30,
500mm tube, 3.56mm outside diameter, 0.41mm wall thickness, 2.74mm inside diameter, as drawn
Expand
View Pricing
GF14617177
Brass alloy, Cu70Zn30,
1m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF74806274
Brass alloy, Cu70Zn30,
10m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF61723526
Brass alloy, Cu70Zn30,
sphere, 3.0mm diameter, 5.08μm sphericity, precision sphere, grade 200, 100pcs
Expand
View Pricing
GF87105012
Brass alloy, Cu70Zn30,
25m wire, 0.6mm diameter, as drawn
Expand
View Pricing
GF29289897
Brass alloy, Cu70Zn30,
1m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF93679532
Brass alloy, Cu70Zn30,
10m wire, 0.6mm diameter, as drawn
Expand
View Pricing
GF51896841
Brass alloy, Cu70Zn30,
5m wire, 0.6mm diameter, as drawn
Expand
View Pricing
GF27147382
Brass alloy, Cu70Zn30,
5m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF31572132
Brass alloy, Cu70Zn30,
50m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF88126689
Brass alloy, Cu70Zn30,
50m wire, 0.6mm diameter, as drawn
Expand
View Pricing
GF90121801
Brass alloy, Cu70Zn30,
10m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF02815174
Brass alloy, Cu70Zn30,
1m wire, 0.6mm diameter, as drawn
Expand
View Pricing
GF93423157
Brass alloy, Cu70Zn30,
25m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF61237223
Brass alloy, Cu70Zn30,
5m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF54652574
Brass alloy, Cu70Zn30,
25m wire, 0.025mm diameter, as drawn
Expand
View Pricing