Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified
Products
Product Category
flasks (20)
Manufacturer
Capacity (mL)
Package Size
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
BR36938
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 5 mL, polypropylene stopper, neck joint: ST/NS 10/19
Expand
View Pricing
BR36983
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 1000 mL, neck joint: ST/NS 24/29, glass stopper
Expand
View Pricing
BR36952
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 500 mL, neck joint: ST/NS 19/26, polypropylene stopper
Expand
View Pricing
BR36950
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 200 mL, neck joint: ST/NS 14/23, polypropylene stopper
Expand
View Pricing
BR36949
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 100 mL, neck joint: ST/NS 14/23, polypropylene stopper
Expand
View Pricing
BR36951
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 250 mL, neck joint: ST/NS 14/23, polypropylene stopper
Expand
View Pricing
BR36943
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 10 mL, neck joint: ST/NS 10/19, polypropylene stopper
Expand
View Pricing
BR36954
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 2000 mL, neck joint: ST/NS 29/32, polypropylene stopper
Expand
View Pricing
BR36981
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 250 mL, neck joint: ST/NS 14/23, glass stopper
Expand
View Pricing
BR36968
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 5 mL, neck joint: ST/NS 10/19, glass stopper
Expand
View Pricing
BR36982
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 500 mL, neck joint: ST/NS 14/23, glass stopper
Expand
View Pricing
BR36979
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 100 mL, neck joint: ST/NS 14/23, glass stopper
Expand
View Pricing
BR36973
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 10 mL, neck joint: ST/NS 10/19, glass stopper
Expand
View Pricing
BR36980
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 200 mL, neck joint: ST/NS 14/23, glass stopper
Expand
View Pricing
BR36953
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
camber 1000 mL, neck joint: ST/NS 24/29, polypropylene stopper
Expand
View Pricing
BR36978
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 50 mL, neck joint: ST/NS 12/21, glass stopper
Expand
View Pricing
BR36948
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 50 mL, neck joint: ST/NS 12/21, polypropylene stopper
Expand
View Pricing
BR36984
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 2000 mL, neck joint: ST/NS 29/32, glass stopper
Expand
View Pricing
BR36947
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 25 mL, neck joint: ST/NS 10/19, polypropylene stopper
Expand
View Pricing
BR36977
BRAND
®
volumetric flask BLAUBRAND
®
USP conformity certified,
capacity 25 mL, neck joint: ST/NS 10/19, glass stopper
Expand
View Pricing