Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
volumetric flask BLAUBRAND
®
class A DE-M marking
Products
Product Category
flasks (3)
Manufacturer
Capacity (mL)
Glassware Class
Package Size
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
BR37268
BRAND
®
volumetric flask BLAUBRAND
®
class A DE-M marking,
capacity 20 mL, neck joint: ST/NS 12/21, glass stopper
Expand
View Pricing
BR37245
BRAND
®
volumetric flask BLAUBRAND
®
class A DE-M marking,
capacity 20 mL, neck joint: ST/NS 12/21, polypropylene stopper
Expand
View Pricing
BR37241
BRAND
®
volumetric flask BLAUBRAND
®
class A DE-M marking,
capacity 10 mL, joint: 7/16, polypropylene stopper
Expand
View Pricing