Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
test tube racks, PP
Products
Product Category
racks (18)
Color
Manufacturer
Height (mm)
Length (mm)
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
Z334146
BRAND
®
test tube racks, PP,
Holds 21 x 30 mm tubes, blue
Expand
View Pricing
Z334243
BRAND
®
test tube racks, PP,
Holds 55 x 16 mm tubes, red
Expand
View Pricing
Z334162
BRAND
®
test tube racks, PP,
Holds 40 x 20 mm tubes, blue
Expand
View Pricing
Z334154
BRAND
®
test tube racks, PP,
Holds 32 x 25 mm tubes, blue
Expand
View Pricing
Z334286
BRAND
®
test tube racks, PP,
Holds 21 x 30 mm tubes, white
Expand
View Pricing
Z334278
BRAND
®
test tube racks, PP,
Holds 84 x 13 mm tubes, red
Expand
View Pricing
Z334219
BRAND
®
test tube racks, PP,
Holds 21 x 30 mm tubes, red
Expand
View Pricing
Z334294
BRAND
®
test tube racks, PP,
Holds 32 x 25 mm tubes, white
Expand
View Pricing
Z334308
BRAND
®
test tube racks, PP,
Holds 40 x 20 mm tubes, white
Expand
View Pricing
Z334170
BRAND
®
test tube racks, PP,
Holds 55 x 16 mm tubes, blue
Expand
View Pricing
Z334316
BRAND
®
test tube racks, PP,
Holds 55 x 16 mm tubes, white
Expand
View Pricing
Z334189
BRAND
®
test tube racks, PP,
Holds 55 x 18 mm tubes, blue
Expand
View Pricing
Z334324
BRAND
®
test tube racks, PP,
Holds 55 x 18 mm tubes, white
Expand
View Pricing
Z334235
BRAND
®
test tube racks, PP,
Holds 40 x 20 mm tubes, red
Expand
View Pricing
Z334227
BRAND
®
test tube racks, PP,
Holds 32 x 25 mm tubes, red
Expand
View Pricing
Z334197
BRAND
®
test tube racks, PP,
Holds 84 x 13 mm tubes, blue
Expand
View Pricing
Z334332
BRAND
®
test tube racks, PP,
Holds 84 x 13 mm tubes, white
Expand
View Pricing
Z334251
BRAND
®
test tube racks, PP,
Holds 55 x 18 mm tubes, red
Expand
View Pricing