Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
PD-Tips II
Products
Product Category
pipette tips (18)
Manufacturer
Volume (ml)
Sterility
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
BR705710
BRAND
®
PD-Tips II,
volume 5 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705718
BRAND
®
PD-Tips II,
volume 50 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705708
BRAND
®
PD-Tips II,
volume 2.5 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705714
BRAND
®
PD-Tips II,
volume 12.5 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705746
BRAND
®
PD-Tips II,
volume 25 mL, BIO-CERT
®
, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705740
BRAND
®
PD-Tips II,
BIO-CERT
®
, volume 5 mL, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705712
BRAND
®
PD-Tips II,
volume 10 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705744
BRAND
®
PD-Tips II,
volume 12.5 mL, BIO-CERT
®
, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705706
BRAND
®
PD-Tips II,
volume 1.25 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705748
BRAND
®
PD-Tips II,
BIO-CERT
®
, volume 50 mL, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705700
BRAND
®
PD-Tips II,
volume 0.1 mL, non-sterile, bulk, polypropylene cylinder, LCP, type encoded
Expand
View Pricing
BR705702
BRAND
®
PD-Tips II,
volume 0.5 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705738
BRAND
®
PD-Tips II,
BIO-CERT
®
, volume 2.5 mL, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705732
BRAND
®
PD-Tips II,
volume 0.5 mL, BIO-CERT
®
, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705734
BRAND
®
PD-Tips II,
BIO-CERT
®
, volume 1 mL, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705742
BRAND
®
PD-Tips II,
volume 10 mL, BIO-CERT
®
, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705736
BRAND
®
PD-Tips II,
BIO-CERT
®
, volume 1.25 mL, individually wrapped, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705730
BRAND
®
PD-Tips II,
BIO-CERT
®
, volume 0.1 mL, individually wrapped, polypropylene cylinder, LCP, type encoded
Expand
View Pricing
BR705716
BRAND
®
PD-Tips II,
volume 25 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
BR705704
BRAND
®
PD-Tips II,
volume 1 mL, non-sterile, bulk, polypropylene cylinder, high-density polyethylene piston, type encoded
Expand
View Pricing
Page 1 of 2
Page 1 of 2