Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern
Products
Product Category
burettes (11)
Manufacturer
Volume (ml)
Glassware Class
Color
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
BR21912
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 25 mL, accuracy: ±0.05 mL, clear borosilicate glass 3.3 (Schellbach lateral needle-valve stopcock PTFE key, grad. in black)
Expand
View Pricing
Z323969
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 50 mL, accuracy: ±0.05 mL, clear borosilicate glass 3.3 (Schellbach lateral needle-valve stopcock PTFE key, grad. in blue)
Expand
View Pricing
BR22523
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 50 mL, accuracy: ±0.05 mL, clear borosilicate glass 3.3 (Schellbach lateral needle-valve stopcock PTFE key, grad. in blue)
Expand
View Pricing
Z323950
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 25 mL, accuracy: ±0.030 mL, clear borosilicate glass 3.3 (Schellbach lateral needle-valve stopcock PTFE key, grad. in blue)
Expand
View Pricing
BR22522
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 25 mL, accuracy: ±0.030 mL, clear borosilicate glass 3.3 (Schellbach lateral needle-valve stopcock PTFE key, grad. in blue)
Expand
View Pricing
BR22318
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 50 mL, accuracy: ±0.10 mL, amber borosilicate glass (lateral needle-valve stopcock PTFE-key, grad. in white)
Expand
View Pricing
BR22316
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 25 mL, accuracy: ±0.08 mL, amber borosilicate glass (lateral needle-valve stopcock PTFE-key, grad. in white)
Expand
View Pricing
BR21913
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 50 mL, accuracy: ±0.08 mL, clear borosilicate glass 3.3 (Schellbach lateral needle-valve stopcock PTFE key, grad. in black)
Expand
View Pricing
BR21911
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 10 mL, accuracy: ±0.030 mL, clear borosilicate glass 3.3 (Schellbach lateral needle-valve stopcock PTFE key, grad. in black)
Expand
View Pricing
Z323942
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 10 mL, accuracy: ±0.020 mL, clear borosilicate glass 3.3 (Schellbach lateral needle-valve stopcock PTFE key, grad. in blue)
Expand
View Pricing
BR22521
BRAND
®
BLAUBRAND
®
automatic burette, Pellet pattern,
volume 10 mL, accuracy: ±0.020 mL, clear borosilicate glass 3.3 (Schellbach lateral needle-valve stopcock PTFE key, grad. in blue)
Expand
View Pricing