Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
, accu-jet
®
S pipette controller
Products
Product Category
pipettors (8)
Manufacturer
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
BR26350
BRAND
®
, accu-jet
®
S pipette controller,
anthracite, AC/DC input 100 - 240 V, 50 - 60 Hz, universal plug set
Expand
View Pricing
BR26351
BRAND
®
, accu-jet
®
S pipette controller,
berry, AC/DC input 100 - 240 V, 50 - 60 Hz, universal plug set
Expand
View Pricing
BR26353
BRAND
®
, accu-jet
®
S pipette controller,
amethyst, AC/DC input 100 - 240 V, 50 - 60 Hz, universal plug set
Expand
View Pricing
BR26352
BRAND
®
, accu-jet
®
S pipette controller,
petrol, AC/DC input 100 - 240 V, 50 - 60 Hz, universal plug set
Expand
View Pricing
BR26360
BRAND
®
, accu-jet
®
S pipette controller,
anthracite, without power adapter
Expand
View Pricing
BR26362
BRAND
®
, accu-jet
®
S pipette controller,
petrol, without power adapter
Expand
View Pricing
BR26361
BRAND
®
, accu-jet
®
S pipette controller,
berry, without power adapter
Expand
View Pricing
BR26363
BRAND
®
, accu-jet
®
S pipette controller,
amethyst, without power adapter
Expand
View Pricing