Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
96-well PCR plate
Products
Product Category
plates (16)
Manufacturer
Binding Type
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
BR781350
BRAND
®
96-well PCR plate,
non-skirted, clear, cut corner H12, standard profile
Expand
View Pricing
BR781368
BRAND
®
96-well PCR plate,
non-skirted, clear, cut corner A12, standard profile
Expand
View Pricing
BR781375
BRAND
®
96-well PCR plate,
semi-skirted, clear, cut corner A12, standard profile
Expand
View Pricing
BR781366
BRAND
®
96-well PCR plate,
non-skirted, clear, cut corner H12, low profile
Expand
View Pricing
BR781357
BRAND
®
96-well PCR plate,
semi-skirted, white, cut corner A1, standard profile
Expand
View Pricing
BR781364
BRAND
®
96-well PCR plate,
semi-skirted, white, cut corner H12
Expand
View Pricing
BR781365
BRAND
®
96-well PCR plate,
semi-skirted, white, cut corner H12, for LightCycler
®
Expand
View Pricing
BR781373
BRAND
®
96-well PCR plate,
semi-skirted, raised skirt, clear, cut corner A1, low profile
Expand
View Pricing
BR781400
BRAND
®
96-well PCR plate,
semi-skirted, clear, cut corner H12, standard profile
Expand
View Pricing
BR781354
BRAND
®
96-well PCR plate,
non-skirted, white, cut corner H12, standard profile
Expand
View Pricing
BR781374
BRAND
®
96-well PCR plate,
semi-skirted (raised skirt), white, cut corner A1, low profile
Expand
View Pricing
BR781372
BRAND
®
96-well PCR plate,
semi-skirted, white, cut corner A12, low profile
Expand
View Pricing
BR781371
BRAND
®
96-well PCR plate,
semi-skirted, clear, cut corner A12, low profile
Expand
View Pricing
BR781376
BRAND
®
96-well PCR plate,
semi-skirted, white, cut corner A12, standard profile
Expand
View Pricing
BR781367
BRAND
®
96-well PCR plate,
non-skirted, white, cut corner H12, low profile
Expand
View Pricing
BR781369
BRAND
®
96-well PCR plate,
non-skirted, white, cut corner A12, standard profile
Expand
View Pricing