
Boc-Thr(t-Bu)-OH

Synonym(s):
Boc-O-tert-butyl-L-threonine, Boc-Thr(tBu)-OH
Linear Formula:
(CH3)3COCH(CH3)CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
275.34
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing