
Boc-propargyl-Gly-OH

Synonym(s):
(S)-2-(Boc-amino)-4-pentynoic acid, Boc-L-2-propargylglycine, Boc-Pra-OH
Empirical Formula (Hill Notation):
C10H15NO4
CAS Number:
Molecular Weight:
213.23
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing