
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate

Synonym(s):
β-DPNH, β-NADH, DPNH, Diphosphopyridine nucleotide, reduced form, NADH
Empirical Formula (Hill Notation):
C21H27N7Na2O14P2 · xH2O
CAS Number:
Molecular Weight:
709.40 (anhydrous basis)
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-4 of 4
Compare
Product Number
Description
Pricing