Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bleomycin sulfate from
Streptomyces verticillus
Synonym(s):
Blenoxane, Bleo, Blexane
CAS Number:
9041-93-4
EC Number:
232-925-2
Products
Product Category
antibiotics (4)
bioactive small molecules (1)
Brand
Sigma-Aldrich (4)
Biological Source
Color
Physical Form
Product Line
Activity Spectrum
Quality Segment
Mode of Action
Special Grade
Biotechnology Performance Certified (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
B5507
Bleomycin sulfate from
Streptomyces verticillus
,
crystalline, 1.5-2.0 U/mg
Expand
View Pricing
B2434
Bleomycin sulfate from
Streptomyces verticillus
,
BioXtra, crystalline
Expand
View Pricing
B8416
Bleomycin sulfate from
Streptomyces verticillus
,
crystalline, suitable for cell culture, BioReagent
Expand
View Pricing
15361
Bleomycin sulfate from
Streptomyces verticillus
,
suitable for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Expand
View Pricing