Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Biohazard disposal bags
Products
Product Category
bags (4)
Manufacturer
Height (mm)
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
HS10320
Biohazard disposal bags,
H × W × thickness 305 mm × 203 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)
Expand
View Pricing
HS10321
Biohazard disposal bags,
H × W × thickness 483 mm × 356 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)
Expand
View Pricing
HS10323
Biohazard disposal bags,
H × W × thickness 838 mm × 635 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)
Expand
View Pricing
HS10322
Biohazard disposal bags,
H × W × thickness 584 mm × 483 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)
Expand
View Pricing