
Bicalutamide Related Compound A

Synonym(s):
N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfinyl]-2-hydroxy-2-methylpropanamide, Bicalutamide sulfoxide
Empirical Formula (Hill Notation):
C18H14F4N2O3S
CAS Number:
Molecular Weight:
414.37
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2