Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Glucuronidase from bovine liver
Synonym(s):
β-D-Glucuronide glucuronosohydrolase
CAS Number:
9001-45-0
EC Number:
232-606-8
Products
Brand
Sigma-Aldrich (2)
Biological Source
Physical Form
Tissue
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G0251
β-Glucuronidase from bovine liver,
Type B-1, ≥1,000,000 units/g solid
Expand
View Pricing
G0376
β-Glucuronidase from bovine liver,
Type B-3, ≥2,000,000 units/g solid
Expand
View Pricing