Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Galactosidase from
Escherichia coli
Synonym(s):
β-D-Galactoside galactohydrolase, Lactase
CAS Number:
9031-11-2
EC Number:
232-864-1
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (5)
Biological Source
Physical Form
Recombinant Host
Chemical Composition
Quality Segment
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
G5635
β-Galactosidase from
Escherichia coli
,
Grade VIII, lyophilized powder, ≥500 units/mg protein
Expand
View Pricing
G3153
β-Galactosidase from
Escherichia coli
,
lyophilized powder, ≥500 units/mg protein
Expand
View Pricing
G6008
β-Galactosidase from
Escherichia coli
,
Grade VI, lyophilized powder, ≥250 units/mg protein
Expand
View Pricing
G4155
β-Galactosidase from
Escherichia coli
,
aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Expand
View Pricing
48275
β-Galactosidase from
Escherichia coli
,
suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Expand
View Pricing