Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benzylamine
Synonym(s):
α-Aminotoluene, Benzylamine
Linear Formula:
C
6
H
5
CH
2
NH
2
CAS Number:
100-46-9
Molecular Weight:
107.15
EC Number:
202-854-1
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
185701
Benzylamine,
ReagentPlus
®
, 99%
Expand
View Pricing
8.01812
Benzylamine,
for synthesis
Expand
View Pricing
407712
Benzylamine,
purified by redistillation, ≥99.5%
Expand
View Pricing
13180
Benzylamine,
for GC derivatization, LiChropur
™
, ≥99.0%
Expand
View Pricing