Skip to Content
Merck
Benzyl alcohol SAJ first grade, ≥98.5%

Benzyl alcohol

Benzyl alcohol SAJ first grade, ≥98.5%
Synonym(s):
Benzenemethanol, Benzyl alcohol
Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-20 of 24
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2