Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benzphetamine hydrochloride
Synonym(s):
N-Benzyl-N,α-dimethylphenethylamine hydrochloride
Empirical Formula (Hill Notation):
C
17
H
21
N · HCl
CAS Number:
5411-22-3
Molecular Weight:
275.82
EC Number:
226-489-2
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1059003
Benzphetamine hydrochloride,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
B8263
Benzphetamine hydrochloride,
Expand
View Pricing