Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bel-Art
®
Switch-Grid
®
Replacement Grids
Products
Product Category
racks (5)
Manufacturer
Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
BAF187453000
Bel-Art
®
Switch-Grid
®
Replacement Grids,
for tubes, 25 ‑ 30 mm, yellow
Expand
View Pricing
BAF187452500
Bel-Art
®
Switch-Grid
®
Replacement Grids,
for tubes, 20 ‑ 25 mm, (Fuchsia)
Expand
View Pricing
BAF187452000
Bel-Art
®
Switch-Grid
®
Replacement Grids,
for tubes, 16 ‑ 20 mm, green
Expand
View Pricing
BAF187451600
Bel-Art
®
Switch-Grid
®
Replacement Grids,
for tubes, 13 ‑ 16 mm, blue
Expand
View Pricing
BAF187451300
Bel-Art
®
Switch-Grid
®
Replacement Grids,
for tubes, 10 ‑ 13 mm, orange
Expand
View Pricing