
(−)-Arctigenin

Synonym(s):
(3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]dihydro-3-[(4-hydroxy-3-methoxyphenyl)methyl]-2(3H)-furanone, (−)-Arctigenin
Empirical Formula (Hill Notation):
C21H24O6
CAS Number:
Molecular Weight:
372.41
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing