
Anti-SMAD7 antibody produced in rabbit

Synonym(s):
Anti-MADH7, Anti-MADH8, Anti-SMAD family member 7
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2