Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Amyloglucosidase from
Aspergillus niger
Synonym(s):
1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase
CAS Number:
9032-08-0
EC Number:
232-877-2
Products
Product Category
enzymes (2)
Brand
Sigma-Aldrich (6)
Sustainability
Biological Source
Color
Feature
Physical Form
pK Value
Chemical Composition
Quality Segment
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
A7095
Amyloglucosidase from
Aspergillus niger
,
≥260 U/mL, aqueous solution
Expand
View Pricing
10115
Amyloglucosidase from
Aspergillus niger
,
lyophilized, powder, ~70 U/mg
Expand
View Pricing
A7420
Amyloglucosidase from
Aspergillus niger
,
lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Expand
View Pricing
10113
Amyloglucosidase from
Aspergillus niger
,
powder, white, ~120 U/mg
Expand
View Pricing
A1602
Amyloglucosidase from
Aspergillus niger
,
ammonium sulfate suspension, ≥40 units/mg protein
Expand
View Pricing
A2910
Amyloglucosidase from
Aspergillus niger
,
Isoelectric focusing marker, pI 3.6
Expand
View Pricing