
Amiodarone Related Compound D

Synonym(s):
(2-Butylbenzofuran-3-yl)(4-hydroxy-3,5-diiodophenyl)methanone, NSC 85437
Empirical Formula (Hill Notation):
C19H16I2O3
CAS Number:
Molecular Weight:
546.14
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2