Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aminopterin
Synonym(s):
(S)-2-{4-[(2,4-Diaminopteridin-6-yl)methylamino]benzamido}pentanedioic acid, 4-Amino-PGA, 4-Aminofolic acid, 4-Aminopteroyl-L-glutamic acid
Empirical Formula (Hill Notation):
C
19
H
20
N
8
O
5
CAS Number:
54-62-6
Molecular Weight:
440.41
EC Number:
200-209-9
Products
Product Category
media supplements (2)
bioactive small molecules (1)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A1784
Aminopterin,
powder
Expand
View Pricing
A5159
Aminopterin,
Hybri-Max
™
, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Expand
View Pricing
A3411
Aminopterin,
powder, BioReagent, suitable for cell culture
Expand
View Pricing