Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum sulfate octadecahydrate
Synonym(s):
Aluminum sulfate hydrate
Linear Formula:
Al
2
(SO
4
)
3
· 18H
2
O
CAS Number:
7784-31-8
Molecular Weight:
666.43
EC Number:
233-135-0
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (2)
Formula Weight
pH Value
Physical Form
Purity
Reagent Type
Core
Special Grade
Puriss (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
227617
Aluminum sulfate octadecahydrate,
≥97%
Expand
View Pricing
11044
Aluminum sulfate octadecahydrate,
puriss., meets analytical specification of Ph. Eur., BP, 100-110%, 51.0-59.0% Al
2
(SO
4
)
3
basis
Expand
View Pricing