Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-magnesium alloy, Al97Mg3
Products
Manufacturer
Physical Form
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 118
Page 1 of 6
Page 1 of 6
Compare
Product Number
Description
Pricing
GF15812557
Aluminum-magnesium alloy, Al97Mg3,
500mm tube, 0.75mm outside diameter, 0.1mm wall thickness, 0.55mm inside diameter, as drawn
Expand
View Pricing
GF48014726
Aluminum-magnesium alloy, Al97Mg3,
foil, 0.5m coil, 0.1mm thickness, as rolled
Expand
View Pricing
GF09201776
Aluminum-magnesium alloy, Al97Mg3,
foil, 200 x 200mm, 0.03mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF25034699
Aluminum-magnesium alloy, Al97Mg3,
200mm tube, 1.0mm outside diameter, 0.075mm wall thickness, 0.85mm inside diameter, as drawn
Expand
View Pricing
GF82917401
Aluminum-magnesium alloy, Al97Mg3,
200mm rod, 20mm diameter
Expand
View Pricing
GF48926111
Aluminum-magnesium alloy, Al97Mg3,
500mm rod, 20mm diameter
Expand
View Pricing
GF34483226
Aluminum-magnesium alloy, Al97Mg3,
foil, 50 x 50mm, 0.03mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF07246302
Aluminum-magnesium alloy, Al97Mg3,
foil, 25 x 25mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF82932217
Aluminum-magnesium alloy, Al97Mg3,
100mm tube, 0.75mm outside diameter, 0.1mm wall thickness, 0.55mm inside diameter, as drawn
Expand
View Pricing
GF99863454
Aluminum-magnesium alloy, Al97Mg3,
1000mm tube, 0.74mm outside diameter, 0.065mm wall thickness, 0.61mm inside diameter, as drawn
Expand
View Pricing
GF82350389
Aluminum-magnesium alloy, Al97Mg3,
foil, 50 x 50mm, 0.1mm thickness, as rolled
Expand
View Pricing
GF93807658
Aluminum-magnesium alloy, Al97Mg3,
1000mm rod, 20mm diameter
Expand
View Pricing
GF67169379
Aluminum-magnesium alloy, Al97Mg3,
1000mm tube, 1.0mm outside diameter, 0.075mm wall thickness, 0.85mm inside diameter, as drawn
Expand
View Pricing
GF66526500
Aluminum-magnesium alloy, Al97Mg3,
200mm tube, 0.75mm outside diameter, 0.1mm wall thickness, 0.55mm inside diameter, as drawn
Expand
View Pricing
GF29556119
Aluminum-magnesium alloy, Al97Mg3,
100mm tube, 0.53mm outside diameter, 0.075mm wall thickness, 0.38mm inside diameter, as drawn
Expand
View Pricing
GF27903821
Aluminum-magnesium alloy, Al97Mg3,
foil, 2m coil, 0.25mm thickness, as rolled
Expand
View Pricing
GF93798411
Aluminum-magnesium alloy, Al97Mg3,
foil, 100 x 100mm, 0.1mm thickness, as rolled
Expand
View Pricing
GF89320743
Aluminum-magnesium alloy, Al97Mg3,
100mm rod, 20mm diameter
Expand
View Pricing
GF34600272
Aluminum-magnesium alloy, Al97Mg3,
100mm tube, 1.0mm outside diameter, 0.075mm wall thickness, 0.85mm inside diameter, as drawn
Expand
View Pricing
GF64798903
Aluminum-magnesium alloy, Al97Mg3,
500mm tube, 0.74mm outside diameter, 0.065mm wall thickness, 0.61mm inside diameter, as drawn
Expand
View Pricing
Page 1 of 6
Page 1 of 6