Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
round-bottom storage flask with stopcock-equipped septum-inlet
Products
Product Category
flasks (7)
Brand
Sigma-Aldrich (7)
Manufacturer
Capacity (mL)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z107298
Aldrich
®
round-bottom storage flask with stopcock-equipped septum-inlet,
with 2 mm glass stopcock, capacity 250 mL, joint: ST/NS 24/40
Expand
View Pricing
Z101982
Aldrich
®
round-bottom storage flask with stopcock-equipped septum-inlet,
capacity 1,000 mL, with 4 mm PTFE stopcock, joint: ST/NS 24/40
Expand
View Pricing
Z107255
Aldrich
®
round-bottom storage flask with stopcock-equipped septum-inlet,
with 2 mm glass stopcock, capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
Z517577
Aldrich
®
round-bottom storage flask with stopcock-equipped septum-inlet,
capacity 250 mL, with 2 mm glass stopcock, joint: ST/NS 29/32
Expand
View Pricing
Z107328
Aldrich
®
round-bottom storage flask with stopcock-equipped septum-inlet,
capacity 1,000 mL, with 4 mm glass stopcock, joint: ST/NS 24/40
Expand
View Pricing
Z103365
Aldrich
®
round-bottom storage flask with stopcock-equipped septum-inlet,
capacity 250 mL, with 2 mm PTFE stopcock, joint: ST/NS 14/20
Expand
View Pricing
Z517615
Aldrich
®
round-bottom storage flask with stopcock-equipped septum-inlet,
with 2 mm PTFE stopcock, capacity 250 mL, joint: ST/NS 29/32
Expand
View Pricing