Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
addition funnels with PTFE needle valve
Products
Product Category
funnels (9)
Brand
Sigma-Aldrich (9)
Capacity (mL)
Height (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z419958
Aldrich
®
addition funnels with PTFE needle valve,
Jacketed, capacity 250 mL, joint: ST/NS 24/40
Expand
View Pricing
Z419931
Aldrich
®
addition funnels with PTFE needle valve,
Jacketed, capacity 100 mL, joint: ST/NS 24/40
Expand
View Pricing
Z419966
Aldrich
®
addition funnels with PTFE needle valve,
Jacketed, capacity 500 mL, joint: ST/NS 24/40
Expand
View Pricing
Z517658
Aldrich
®
addition funnels with PTFE needle valve,
Jacketed, capacity 100 mL, joint: ST/NS 29/32
Expand
View Pricing
Z517666
Aldrich
®
addition funnels with PTFE needle valve,
Jacketed, capacity 250 mL, joint: ST/NS 29/32
Expand
View Pricing
Z517682
Aldrich
®
addition funnels with PTFE needle valve,
Unjacketed, capacity 100 mL, joint: ST/NS 29/32
Expand
View Pricing
Z517674
Aldrich
®
addition funnels with PTFE needle valve,
Jacketed, capacity 500 mL, joint: ST/NS 29/32
Expand
View Pricing
Z419974
Aldrich
®
addition funnels with PTFE needle valve,
Unjacketed, capacity 100 mL, joint: ST/NS 24/40
Expand
View Pricing
Z419982
Aldrich
®
addition funnels with PTFE needle valve,
Unjacketed, capacity 250 mL, joint: ST/NS 24/40
Expand
View Pricing