Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
addition funnel
Products
Product Category
funnels (10)
Brand
Sigma-Aldrich (10)
Capacity (mL)
Height (mm)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z122467
Aldrich
®
addition funnel,
capacity 50 mL, joint: ST/NS 14/20
Expand
View Pricing
Z548650
Aldrich
®
addition funnel,
capacity 250 mL, joint: ST/NS 24/40
Expand
View Pricing
Z548634
Aldrich
®
addition funnel,
capacity 100 mL, joint: ST/NS 24/40
Expand
View Pricing
Z548669
Aldrich
®
addition funnel,
capacity 250 mL, joint: ST/NS 29/32
Expand
View Pricing
Z548626
Aldrich
®
addition funnel,
capacity 100 mL, joint: ST/NS 14/20
Expand
View Pricing
Z548618
Aldrich
®
addition funnel,
capacity 25 mL, joint: ST/NS 14/20
Expand
View Pricing
Z548642
Aldrich
®
addition funnel,
capacity 100 mL, joint: ST/NS 29/32
Expand
View Pricing
Z548596
Aldrich
®
addition funnel,
capacity 10 mL, joint: ST/NS 14/20
Expand
View Pricing
Z548677
Aldrich
®
addition funnel,
capacity 500 mL, joint: ST/NS 24/40
Expand
View Pricing
Z548685
Aldrich
®
addition funnel,
capacity 500 mL, joint: ST/NS 29/32
Expand
View Pricing