
α-Hydroxyisobutyronitrile β-D-glucopyranoside

Synonym(s):
α-Hydroxyisobutyronitrile β-D-glucose, 2-(β-D-Glucopyranosyloxy)-2-methylpropionitrile, Linamarin
Empirical Formula (Hill Notation):
C10H17NO6
CAS Number:
Molecular Weight:
247.25
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing