Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
S-4x Universal Large Rotor
Products
Product Category
adapters (19)
Manufacturer
Diameter (mm)
Height (mm)
Package Size
Technique
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
EP5920737007
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, MTP, DWP, PCRplates, pack of 2
Expand
View Pricing
EP5920750003
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, 1 x 250 mL Corning
®
bottle, pack of 2
Expand
View Pricing
EP5920744003
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, 1 x 500 mL Corning
®
bottle, pack of 2
Expand
View Pricing
EP5910751001
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
for 50 mL conical tubes/microtiter plates/250 mL flat-bottom or 175-225 mL conical tubes/microtiter plates, pkg of 2 ea
Expand
View Pricing
EP5920751000
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, 24 x 14 mL tubes, pack of 2
Expand
View Pricing
EP5910760000
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
for 500 mL conical tubes, pack of 2 ea
Expand
View Pricing
EP5920747002
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, 92 x 1.5/2.0 mL tube, pack of 2
Expand
View Pricing
EP5920740008
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
Holds 250 mL flat bottom bottles, pack of 2
Expand
View Pricing
EP5920745000
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, 1 x 500 mL Nalgene
®
bottle, pack of 2
Expand
View Pricing
EP5910752008
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
for 5 mL and 15 mL conical tubes/microtiter and deepwell plates, pkg of 2 ea
Expand
View Pricing
EP5920742000
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, 52 x 5 mL FACS, pack of 2
Expand
View Pricing
EP5920741004
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
Holds 750 mL flat bottom bottles, pack of 2
Expand
View Pricing
EP5910757000
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
for 750 mL conical tubes, pkg of 2 ea
Expand
View Pricing
EP5920739000
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, 49 x 2.6 - 8 mL tubes, pack of 2
Expand
View Pricing
EP5920748009
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, 7 x 50 mL conical tubes, pack of 2
Expand
View Pricing
EP5910762003
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
fpr 17.5x100 mm round-bottom tubes, pack of 2 ea
Expand
View Pricing
EP5910755007
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
13x75-100 mm round-bottom rubes, pkg of 2 ea
Expand
View Pricing
EP5910754000
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
for 16x75-100 mm round-bottom tubes, pkg of 2 ea
Expand
View Pricing
EP5920738003
Adapter for Eppendorf
®
S-4x Universal Large Rotor,
capacity, 37 x 5.5 - 12 mL tubes, pack of 2
Expand
View Pricing