Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
S-4x1000 Rotor
Products
Product Category
adapters (16)
Manufacturer
Diameter (mm)
Height (mm)
Package Size
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
EP5920717006
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 2 bottle 250 mL flat, 175 - 225/250 mL conical, for use with high-capacity buckets
Expand
View Pricing
EP5920711008
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 7 x 50 mL Conical tubes, for use with medium-capacity buckets, pack of 2
Expand
View Pricing
EP5920700006
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 1 Liter Nalgene, pack of 2
Expand
View Pricing
EP5920710001
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 22 x Eppendorf Tube
®
5.0 mL, for use with medium-capacity buckets, pack of 2
Expand
View Pricing
EP5920715003
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 12 x 50 mL conical tubes, for use with high-capacity buckets, pack of 2
Expand
View Pricing
EP5920707000
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 33 x 5.5 - 12 mL tubes, for use with medium-capacity buckets, pack of 2
Expand
View Pricing
EP5920712004
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 16 x Eppendorf Tube
®
5.0 mL, for use with medium-capacity buckets, pack of 2
Expand
View Pricing
EP5920706004
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 35 x 2.6 - 8 mL conical tubes, for use with medium-capacity buckets, pack of 2
Expand
View Pricing
EP5920718002
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 49 x 2.6 - 8 mL tubes, for use with high-capacity buckets, pack of 2
Expand
View Pricing
EP5920701002
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 1 x 600 mL Nalgene
®
bottle, pack of 2
Expand
View Pricing
EP5920722000
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 29 x 9 mL tubes, for use with high-capacity buckets, pack of 2
Expand
View Pricing
EP5920716000
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 27 x Eppendorf Tube
®
5.0 mL, for use with high-capacity buckets, pack of 2
Expand
View Pricing
EP5920724002
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 52 x 5 mL FACS, for use with high-capacity buckets, pack of 2
Expand
View Pricing
EP5920720007
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 36 x 5.5 - 12 mL tubes, for use with high-capacity buckets, pack of 2
Expand
View Pricing
EP5920708007
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 28 x 9 mL tubes, for use with medium-capacity buckets, pack of 2
Expand
View Pricing
EP5920703005
Adapter for Eppendorf
®
S-4x1000 Rotor,
capacity, 1 x 500 mL Nalgene
®
bottle, pack of 2
Expand
View Pricing