Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
FA-45-6-30 Rotor
Products
Product Category
adapters (11)
Manufacturer
Diameter (mm)
Package Size
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
EP5820717009
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 x 15 mL conical tube, pack of 2
Expand
View Pricing
EP5820720000
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 x 15–18 mL tube, pack of 2
Expand
View Pricing
EP5820726008
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 2.6 - 5 mL round-bottom tubes, pack of 2
Expand
View Pricing
EP5820730005
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 Eppendorf Tube
®
5.0 mL, pack of 2
Expand
View Pricing
EP5820721006
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 x 20–30 mL tube, pack of 2
Expand
View Pricing
EP5820727004
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 x 7.5 - 12 mL tube, pack of 2
Expand
View Pricing
EP5820722002
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 x 35 mL tube, pack of 2
Expand
View Pricing
EP5820729007
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 x 7 - 17 mL tube, pack of 2
Expand
View Pricing
EP5820728000
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 x 5.5 - 10 mL tube, pack of 2
Expand
View Pricing
EP5820725001
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 x 4 - 8 mL round bottom tubes, pack of 2
Expand
View Pricing
EP5820719001
Adapter for Eppendorf
®
FA-45-6-30 Rotor,
capacity, 1 x 10 mL Oak Ridge tubes, pack of 2
Expand
View Pricing