Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Acetic acid-d
4
Synonym(s):
Acetic-d
3
acid-d, Tetradeuteroacetic acid
Linear Formula:
CD
3
CO
2
D
CAS Number:
1186-52-3
Molecular Weight:
64.08
EC Number:
214-693-4
Products
Product Category
solvents (3)
stable isotopes (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
151785
Acetic acid-d
4
,
≥99.5 atom % D
Expand
View Pricing
233315
Acetic acid-d
4
,
≥99.9 atom % D
Expand
View Pricing
416886
Acetic acid-d
4
,
≥99.5 atom % D, contains 0.03 % (v/v) TMS
Expand
View Pricing