Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ACES
Synonym(s):
N
-(2-Acetamido)-2-aminoethanesulfonic acid,
N
-(Carbamoylmethyl)-2-aminoethanesulfonic acid,
N
-(Carbamoylmethyl)taurine
Linear Formula:
H
2
NCOCH
2
NHCH
2
CH
2
SO
3
H
CAS Number:
7365-82-4
Molecular Weight:
182.20
EC Number:
230-908-4
Products
Product Category
buffers (5)
Brand
Sigma-Aldrich (5)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Special Grade
Anhydrous (1)
BioPerformance Certified (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
A9758
ACES,
≥99.0% (titration)
Expand
View Pricing
A3594
ACES,
BioPerformance Certified, ≥99.0%, suitable for cell culture
Expand
View Pricing
A7949
ACES,
BioXtra, ≥99.0% (titration)
Expand
View Pricing
RDD010
ACES,
anhydrous, free-flowing, Redi-Dri
™
, ≥99.0%
Expand
View Pricing
00194
ACES,
BioUltra, ≥99.5% (T)
Expand
View Pricing