Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
α,α,α-Trifluorotoluene solution
Synonym(s):
Benzotrifluoride
Linear Formula:
C
6
H
5
CF
3
CAS Number:
98-08-8
Molecular Weight:
146.11
EC Number:
214-061-8
Products
Product Category
stable isotopes (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
551406
α,α,α-Trifluorotoluene solution,
NMR reference standard, 0.05% in benzene-d
6
(99.6 atom % D)
Expand
View Pricing
612685
α,α,α-Trifluorotoluene solution,
NMR reference standard, 0.05% in benzene-d
6
(99.6 atom % D), NMR tube size 3 mm × 8 in.
Expand
View Pricing
733806
α,α,α-Trifluorotoluene solution,
NMR reference standard, 0.05% in benzene-d
6
(99.6 atom % D), NMR tube size 10 mm × 8 in.
Expand
View Pricing