
6-Bromo-2-naphthyl β-D-glucopyranoside

Synonym(s):
6-Bromo-2-naphthyl β-D-glucoside
Empirical Formula (Hill Notation):
C16H17BrO6
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing