
5-(Ethylthio)-1H-tetrazole

Empirical Formula (Hill Notation):
C3H6N4S
CAS Number:
Molecular Weight:
130.17
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing