Skip to Content
Merck
4-Fluorophenylboronic acid ≥95%

4-Fluorophenylboronic acid

4-Fluorophenylboronic acid ≥95%
Synonym(s):
(4-Fluorophenyl)boric acid, (4-Fluorophenyl)dihydroxyborane, (4-Fluorophenyl)dihydroxyboron, (p-Fluorophenyl)boric acid, 4-Fluorobenzeneboronic acid, p-Fluorobenzylboronic acid, p-Fluorophenylboronic acid, NSC 142683
Linear Formula:
FC6H4B(OH)2
CAS Number:
Molecular Weight:
139.92

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing