Skip to Content
Merck
4-(Dimethylamino)phenyldiphenylphosphine 95%

4-(Dimethylamino)phenyldiphenylphosphine

4-(Dimethylamino)phenyldiphenylphosphine 95%
Synonym(s):
4-(Dimethylamino)triphenylphosphine, 4-(Diphenylphosphino)-N,N-dimethyl-benzenamine, 4-(Diphenylphosphino)-N,N-dimethylaniline
Linear Formula:
(CH3)2NC6H4P(C6H5)2
CAS Number:
Molecular Weight:
305.35

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing