Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-(Dimethylamino)benzaldehyde
Synonym(s):
Ehrlich’s reagent, 4-(Dimethylamino)benzaldehyde
Linear Formula:
(CH
3
)
2
NC
6
H
4
CHO
CAS Number:
100-10-7
Molecular Weight:
149.19
EC Number:
202-819-0
Products
Product Category
building blocks (5)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Puriss P.A. (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
109762
4-(Dimethylamino)benzaldehyde,
98%
Expand
View Pricing
D2004
4-(Dimethylamino)benzaldehyde,
suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
Expand
View Pricing
1.03058
4-(Dimethylamino)benzaldehyde,
GR for analysis Reag. Ph Eur
Expand
View Pricing
8.03057
4-(Dimethylamino)benzaldehyde,
for synthesis
Expand
View Pricing
156477
4-(Dimethylamino)benzaldehyde,
ACS reagent, 99%
Expand
View Pricing
07-4130
4-(Dimethylamino)benzaldehyde,
JIS special grade, ≥99.0%
Expand
View Pricing
33130
4-(Dimethylamino)benzaldehyde,
puriss. p.a., reag. Ph. Eur., ≥99% (perchloric acid titration)
Expand
View Pricing