
4-Chlororesorcinol

Synonym(s):
1,3-Dihydroxy-4-chlorobenzene
Linear Formula:
ClC6H3(OH)2
CAS Number:
Molecular Weight:
144.56
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing