Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Chlorophenoxyacetic acid
Synonym(s):
4-CPA
Empirical Formula (Hill Notation):
C
8
H
7
ClO
3
CAS Number:
122-88-3
Molecular Weight:
186.59
EC Number:
204-581-3
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Physical Form
Product Line
Purity
Functional Group
Application
Markush Class
Markush Group
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
25890
4-Chlorophenoxyacetic acid,
≥98.0% (T)
Expand
View Pricing
C0413
4-Chlorophenoxyacetic acid,
BioReagent, suitable for plant cell culture, crystalline
Expand
View Pricing