![4′-Chlorodiazepam ≥98% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/207/117/f0ae67d4-6571-4572-a7f9-122a7f380faa/640/f0ae67d4-6571-4572-a7f9-122a7f380faa.png)
4′-Chlorodiazepam
![4′-Chlorodiazepam ≥98% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/207/117/f0ae67d4-6571-4572-a7f9-122a7f380faa/640/f0ae67d4-6571-4572-a7f9-122a7f380faa.png)
Synonym(s):
7-Chloro-5-(4-chlorophenyl)-1,3-dihydro-1-methyl-2H-1,4-benzodiazepin-2-one, Ro 5-4864, Ro5-4864
Empirical Formula (Hill Notation):
C16H12Cl2N2O
CAS Number:
Molecular Weight:
319.19
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing