
3,5-Dichloro-2-hydroxybenzenesulfonyl chloride

Synonym(s):
2,4-Dichlorophenol-6-sulfonyl chloride
Linear Formula:
Cl2C6H2(OH)SO2Cl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing