Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3,3′,5-Triiodo-L-thyronine sodium salt
Synonym(s):
Liothyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt, T
3
Empirical Formula (Hill Notation):
C
15
H
11
I
3
NNaO
4
CAS Number:
55-06-1
Molecular Weight:
672.96
EC Number:
200-223-5
Products
Product Category
hormones (2)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T6397
3,3′,5-Triiodo-L-thyronine sodium salt,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
T5516
3,3′,5-Triiodo-L-thyronine sodium salt,
γ-irradiated, powder, suitable for, suitable for cell culture
Expand
View Pricing